Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Store vial at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4° C as an undiluted liquid. Dilute only prior to immediate use.
This affinity purified antibody is directed against rat VAChT. A BLAST analysis was used to suggest cross-reactivity with the antigen based on 100% homology with the immunizing sequence to mouse and human.
This gene is a member of the vesicular amine transporter family. The encoded transmembrane protein transports acetylcholine into secretory vesicles for release into the extracellular space. Acetylcholine transport utilizes a proton gradient established by a vacuolar ATPase. This gene is located within the first intron of the choline acetyltransferase gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC12716; solute carrier family 18 (vesicular acetylcholine transporter), member 3; solute carrier family 18 (vesicular acetylcholine), member 3; solute carrier family 18 (vesicular monoamine) member 3; Solute carrier family 18 member 3; solute carrier family 18, member 3; VAChT; Vesicular acetylcholine transporter
Gene Aliases: rVAT; SLC18A3; VACHT; Vat
UniProt ID: (Human) Q16572, (Rat) Q62666
Entrez Gene ID: (Human) 6572, (Rat) 60422
Molecular Function:
secondary carrier transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support