Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute in 500 µL of sterile water. Centrifuge to remove any insoluble material.
Specificity of this antibody: VAMP1.
Synapotobrevins, syntaxins, and the synaptosomal-associated protein SNAP25 are the main components of a protein complex involved in the docking and/or fusion of synaptic vesicles with the presynaptic membrane. The protein encoded by this gene is a member of the vesicle-associated membrane protein (VAMP)/synaptobrevin family. Mutations in this gene are associated with autosomal dominant spastic ataxia 1. Multiple alternative splice variants have been described, but the full-length nature of some variants has not been defined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp686H12131; MRXSYP; synaptobrevin 1; Synaptobrevin-1; vamp; VAMP-1; Vesicle-associated membrane protein 1
Gene Aliases: lew; Syb-1; Syb1; VAMP-1; Vamp1
UniProt ID: (Mouse) Q62442, (Rat) Q63666
Entrez Gene ID: (Mouse) 22317, (Rat) 25624
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support