Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Antibody detects endogenous levels of total VAMP 1/2/3.
Synapotobrevins, syntaxins, and the synaptosomal-associated protein SNAP25 are the main components of a protein complex involved in the docking and/or fusion of synaptic vesicles with the presynaptic membrane. The protein encoded by this gene is a member of the vesicle-associated membrane protein (VAMP)/synaptobrevin family. Mutations in this gene are associated with autosomal dominant spastic ataxia 1. Multiple alternative splice variants have been described, but the full-length nature of some variants has not been defined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CEB; Cellubrevin; synaptobrevin 1; synaptobrevin 2; Synaptobrevin 2 (vesicle-associated membrane protein VAMP-2); synaptobrevin II; Synaptobrevin-1; Synaptobrevin-2; Synaptobrevin-3; VAMP-1; VAMP-2; VAMP-3; vesicle-associated membrane 2; Vesicle-associated membrane protein (synaptobrevin 2); Vesicle-associated membrane protein 1; vesicle-associated membrane protein 1 (synaptobrevin 1); Vesicle-associated membrane protein 2; vesicle-associated membrane protein 2 (synaptobrevin 2); Vesicle-associated membrane protein 3
Gene Aliases: CEB; D130027G05Rik; lew; RATVAMPB; RATVAMPIR; SPAX1; SYB; Syb-1; Syb-2; SYB1; SYB2; SYB3; sybII; VAMP-1; VAMP-2; VAMP-3; VAMP1; VAMP2; VAMP3
UniProt ID: (Human) P23763, (Human) P63027, (Human) Q15836, (Mouse) Q62442, (Rat) Q63666, (Mouse) P63044, (Rat) P63045, (Mouse) P63024, (Rat) P63025
Entrez Gene ID: (Human) 6843, (Human) 6844, (Human) 9341, (Mouse) 22317, (Rat) 25624, (Mouse) 22318, (Rat) 24803, (Mouse) 22319, (Rat) 29528
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support