Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Van Gogh is an alternate name for the Drosophila strabismus protein. Van Gogh like-2 (VANGL2) is a 521 amino acidprotein in vertebrates similar to strabismus 1 of Drosophila and a component of the frizzled-disheveled tissue polarity pathway. It interacts through it's C-terminal region with the N-terminal half of DVL1, DVL2 and DVL3 and is a potent tumor suppressor. It ispredominantly involved in the control of early morphogenesis and patterning of both axial midline structures and the development of neural plate. It plays a crucial role in the regulation of planar cell polarity, particularly in the orientation of stereociliary bundles in the cochlea. VANGL2 is required for polarization and movement of myocardializing cells and seems to act via RHOA signaling to regulate this process. It is expressed in a wide range of tissues.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: loop tail associated protein; Loop-tail protein 1; Loop-tail protein 1 homolog; Loop-tail-associated protein; MGC119403; MGC119404; Strabismus 1; Van Gogh-like protein 2; vang; vang (van gogh)-like 2; vang-like 2 (van gogh, Drosophila); Vang-like protein 2
Gene Aliases:
C530001F03Rik; KIAA1215; loop-tail; Lp; LPP1; LTAP; ska17; ska
UniProt ID: (Human) Q9ULK5, (Mouse) Q91ZD4
Entrez Gene ID: (Human) 57216, (Mouse) 93840
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support