Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: NPFNCVCPLS WFGPWVRESH VTLASPEETR CHFPPKNAGR LLLELDYADF GC
VASN may act as an inhibitor of TGF-beta signaling.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein slit-like 2; slit-like 2; Vasorin
Gene Aliases: SLITL2; UNQ314/PRO357/PRO1282; VASN
UniProt ID: (Human) Q6EMK4
Entrez Gene ID: (Human) 114990
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support