Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: DLFDVRDFG KVISAVSRLS LHSIAQNKGI RPFPSEETTE NDDDVYRSLE ELADEHDLGE DIYDCVPCED GGDDIYEDII KVEVQQPMKM GMTEDDKRNC CLLEIQETEA KYYRTLEDIE KNYMSPLRLV LSPADMAAVF INLEDLIKVH HSFLRAIDVS VMVGGSTLAK VFLDFKERLL IYGEYCSHME HAQNTLNQLL ASREDFRQKV EECTLKVQDG KFKLQDLLVV (98-326 aa encoded by BC132967)
VAV2 is the second member of the VAV guanine nucleotide exchange factor family of oncogenes. Unlike VAV1, which is expressed exclusively in hematopoietic cells, VAV2 transcripts were found in most tissues. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Guanine nucleotide exchange factor VAV2; Oncogene VAV2; vav 2 guanine nucleotide exchange factor; vav 2 oncogene; VAV-2; Vav2 oncogene
Gene Aliases: 2810040F13Rik; AI847175; VAV-2; VAV2
UniProt ID: (Human) P52735, (Mouse) Q60992
Entrez Gene ID: (Human) 7410, (Rat) 296603, (Mouse) 22325
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support