Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes a homeobox protein and is almost exclusively expressed in the ventral portion of the retina during development. In mouse studies, this gene was found to be required for the correct formation of the optic fissure and other aspects of retinal development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ventral anterior homeobox 2; ventral anterior homeobox containing gene 2; Ventral retina homeodomain protein
Gene Aliases: DRES93; VAX2; Vex
UniProt ID: (Human) Q9UIW0, (Mouse) Q9WTP9, (Rat) Q9JLZ9
Entrez Gene ID: (Human) 25806, (Mouse) 24113, (Rat) 64572
Molecular Function:
homeodomain transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support