Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: RRAESGGGGGR CGDRSGAGDL RADGGGHSPT EVAGTSASSP AGSRESGADS DGQPGPGEAD HCRRILVRDA KGTIREIVLP KGLDLDRPKR T
Highest antigen sequence identity to the following orthologs - mouse 79%, rat 78%.
This gene encodes a homeobox protein and is almost exclusively expressed in the ventral portion of the retina during development. In mouse studies, this gene was found to be required for the correct formation of the optic fissure and other aspects of retinal development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ventral anterior homeobox 2
Gene Aliases: DRES93; VAX2
UniProt ID: (Human) Q9UIW0
Entrez Gene ID: (Human) 25806
Molecular Function:
homeodomain transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support