Invitrogen
MA1-34975 detects VCAM1 from human samples.
MA1-34975 has been successfully used in ELISA, Flow cytometry, and immunoprecipitation applications.
The MA1-34975 immunogen is: Full length protein (Human)
VCAM-1 is a cell surface sialoglycoprotein expressed by cytokine activated endothelium. The protein has a number of functions including the regulation of leukocyte migration, leukocyte-endothelial cell adhesion and signal transduction and may play a role in a number of inflammatory diseases.