Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: GMTPSKGVL FYGPPGCGKT LLAKAIANEC QANFISIKGP ELLTMWFGES EANVREIFDK ARQAAPCVLF FDELDSIAKA RGGNIGDGGG AADRVINQIL TEMDGMSTKK NVFIIGATNR PDIIDPAILR PGRLDQLIYI PLPDEKSRVA ILKANLRKSP VAKDVDLEFL AKMTNGFSGA DLTEICQRAC KLAIRESIES EIRRERERQT NPSAMEVEED DPVPEIRRDH FEEAMRFARR SVSDNDIRKY EMFAQTLQQS RGFGSFRFPS GNQGGAGPSQ GSGGGTGGSV YTEDNDDDLY G (507-806 aa encoded by BC007562)
The protein encoded by this gene is a member of a family that includes putative ATP-binding proteins involved in vesicle transport and fusion, 26S proteasome function, and assembly of peroxisomes. This protein, as a structural protein, is associated with clathrin, and heat-shock protein Hsc70, to form a complex. It has been implicated in a number of cellular events that are regulated during mitosis, including homotypic membrane fusion, spindle pole body function, and ubiquitin-dependent protein degradation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 15S Mg(2+)-ATPase p97 subunit; AAA ATPase p97; epididymis luminal protein 220; epididymis secretory protein Li 70; homolog of yeast cdc48; Inv protein; or HEL-S-70; p97 atpase; TER ATPase; Transitional endoplasmic reticulum ATPase; Valosin-containing protein; VCP; yeast Cdc48p homolog
Gene Aliases: 3110001E05; ALS14; CDC48; CMT2Y; HEL-220; HEL-S-70; IBMPFD; IBMPFD1; p97; p97/VCP; TERA; VCP
UniProt ID: (Human) P55072, (Mouse) Q01853
Entrez Gene ID: (Human) 7415, (Mouse) 269523
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support