Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
This gene encodes a member of the voltage-dependent anion channel pore-forming family of proteins that are considered the main pathway for metabolite diffusion across the mitochondrial outer membrane. The encoded protein is also thought to be involved in the mitochondrial apoptotic pathway via regulation of BCL2-antagonist/killer 1 protein activity. Pseudogenes have been identified on chromosomes 1, 2, 12 and 21, and alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B36-VDAC; FLJ23841; Outer mitochondrial membrane protein porin 2; RP11-375G3.1; VDAC-2; VDAC-6; voltage-dependent anion channel 2; Voltage-dependent anion-selective channel protein 2; Voltage-dependent anion-selective channel protein 6
Gene Aliases: mVDAC2; mVDAC6; POR; VDAC2; Vdac6
UniProt ID: (Human) P45880, (Mouse) Q60930, (Rat) P81155
Entrez Gene ID: (Human) 7417, (Mouse) 22334, (Rat) 83531
Molecular Function:
voltage-gated ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support