Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Cross-reactivity in ELISA with Mouse VEGF164. No cross-reactivity in ELISA with Human VEGF165, Human VEGF121, Human VEGFc, and Human VEGFd.
VEGF-164 is a gene target that encodes for the vascular endothelial growth factor. This protein is responsible for promoting angiogenesis. VEGF-164 has been shown to be involved in the development of various diseases, including cancer, cardiovascular disease, and diabetic retinopathy. Targeting this gene has become an important strategy in the development of new therapeutics for these conditions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Vascular endothelial growth factor A; Vascular permeability factor; VEGF-A; VPF
Gene Aliases: Vegf; VEGF-A; VEGF164; Vegfa; VPF
UniProt ID: (Rat) P16612
Entrez Gene ID: (Rat) 83785
Molecular Function:
growth factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support