Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive Samples: HT-29, A-549, NCI-H460, H9C2, Mouse lung, Mouse heart, Rat heart
Immunogen sequence: YSIIRRSIQI PEEDRCSHSK KLCPIDMLWD SNKCKCVLQE ENPLAGTEDH SHLQEPALCG PHMMFDEDRC ECVCKTPCPK DLIQHPKNCS CFECKESLET C
The protein encoded by this gene is a member of the platelet-derived growth factor/vascular endothelial growth factor (PDGF/VEGF) family and is active in angiogenesis, lymphangiogenesis, and endothelial cell growth. This secreted protein undergoes a complex proteolytic maturation, generating multiple processed forms which bind and activate VEGFR-2 and VEGFR-3 receptors. This protein is structurally and functionally similar to vascular endothelial growth factor C.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: c-fos induced growth factor (vascular endothelial growth factor D); c-Fos-induced growth factor; FIGF; Vascular endothelial growth factor D; VEGF-D; VEGF4
Gene Aliases: AI325264; FIGF; VEGF-D; VEGFD
UniProt ID: (Human) O43915, (Mouse) P97946, (Rat) O35251
Entrez Gene ID: (Human) 2277, (Mouse) 14205, (Rat) 360457
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support