Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MRLSSSPPRG PQQLSSFGSV DWLSQSSCSG PTHTPRPADF SLGSLPGPGQ TSGAREPPQA VSIKEAAGSS NLPAPERTMA GLSKEPNTLR APRVRTAFTM EQVRTLEGVF QHHQYLSPLE RKRLAREMQL SEVQIKTWFQ NRRMKHKRQM QDPQLHSPFS GSLHAPPAFY STSSGLANGL QLLCPWAPLS GPQALMLPPG SFWGLCQVAQ EALASAGASC CGQPLASHPP TPGRPSLGPA LSTGPRGLCA MPQTGDAF
VENTX is a member of the Lymphoid-enhancing factor/T-cell factors (LEF1/TCF), which are a high-mobility group of transcriptional factors that play essential roles in cell fate determination during early embryogenesis and ontogenesis. VENTX is a member of the Vent family of homeodomain proteins. It is a novel LEF/TCF-associated transcriptional modulator. VENTX is predominately expressed in hematopoietic cells. VENTX may function as a transcriptional repressor and be involved in mesodermal patterning and hemapoietic stem cell maintenance, and has been suggested to have a potential role in the hematopoietic malignancies.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hemopoietic progenitor homeobox protein VENTX2; Homeobox protein VENTX; VENT homeobox homolog; VENT-like homeobox 2; VENT-like homeobox protein 2
Gene Aliases: HPX42B; NA88A; VENTX; VENTX2
UniProt ID: (Human) O95231
Entrez Gene ID: (Human) 27287
Molecular Function:
homeodomain transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support