Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MEEMKKTAIR LPKGKQKPIK TEWNSRCVLF TYFQGDISSV VDEHFSRALS NIKSPQELTP SSQSEGVMLK NDDSISPNQW RYSSPWTKPQ PEVPVTNRAA NCNLHVPGPM AVNQFSPSLA RRASVRPGEL WHFSSLAGTS SLEPGYSHPF PARHLVPEPQ PDGKREPLLS LLQQDRCLAR PQESAARENG NPGQIAGSTG LLFNLPPGSV HYKKLYVSRG SASTSLPNET LSELETPGKY SLTPPNHWGH PHRYLQHL
May act as a specific coactivator for the mammalian TEFs.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein TONDU; TONDU; Transcription cofactor vestigial-like protein 1; vestigial like 1; vestigial-like family member 1; Vgl-1; WUGSC:H_GS188P18.1b
Gene Aliases: TDU; VGL1; VGLL1
UniProt ID: (Human) Q99990
Entrez Gene ID: (Human) 51442
Molecular Function:
transcription cofactor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support