Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 17
Keep as concentrated solution.
Predicted reactivity: Human (96%), Mouse (100%), Rat (98%), Bovine (100%).
Positive Control: mouse brain, rat brain.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
A Na(+)-dependent inorganic phosphate cotransporter; may be involved in neuronal Na+ transport.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Brain-specific Na(+)-dependent inorganic phosphate cotransporter; brain-specific Na-dependent inorganic phosphate cotransporter; SLC17A1; SLC17A1/VGLUT1; solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), member 7; Solute carrier family 17 member 7; Vesicular glutamate transporter 1; VGlU1; VGLUT 1; VGluT1
Gene Aliases: 2900052E22Rik; AI851913; BNPI; Slc17a7; Vglut1
UniProt ID: (Rat) Q62634, (Mouse) Q3TXX4
Entrez Gene ID: (Rat) 116638, (Mouse) 72961
Molecular Function:
secondary carrier transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support