Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
The peptide is homologous in rat and mouse.
Specificity of this antibody: SLC32A1.
The Vesicular GABA Amino Acid Transporter (VGAT) is responsible for transport of the inhibitory neurotransmitter into synaptic vesicles (McIntire et al., 1997). The VGAT protein (also known as the Vesicular Inhibitory Amino Aid Transporter or VIAAT) is expressed in synaptic vesicles of both glycine and GABAergic synapses throughout the CNS (Chaudhry et al., 1998). Expression of the VGAT protein changes during development and also in response to patterns of neuronal activity (De et al., 2005).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GABA and glycine transporter; GVAT; hVIAAT; mVGAT; mVIAAT; rat UNC-47 homolog; rGVAT; RUNC-47; solute carrier family 32 (GABA vesicular transporter), member 1; Solute carrier family 32 member 1; vasicular inhibitory amino acid transporter 10D; Vesicular GABA and glycine transporter; Vesicular GABA transporter; Vesicular inhibitory amino acid transporter
Gene Aliases: R75019; SLC32A1; VGAT; VIAAT
UniProt ID: (Human) Q9H598, (Rat) O35458, (Mouse) O35633
Entrez Gene ID: (Human) 140679, (Rat) 83612, (Mouse) 22348
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support