Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: AARLQEECD YVQMIEVQHK QCLEEAQLEN ETIGCSKMWD NLTCWPATPR GQVVVLACPL IFKLFSSIQG RNVSRSCTDE GWTHLEPGPY PIACGLDDKA ASLDEQQTMF YGSVKTGY (30-146 aa encoded by BC064424 )
The Vasoactive Intestinal Polypeptide Receptor 1 (VPAC1) is a member of a family of three G-protein coupled receptors which mediate VIP (vasoactive intestinal peptide) and PACAP (pituitary adenlyate cyclase activating peptide) function in a variety of tissues. Together, the VPAC1 and VPAC2 receptors compose a subtype of the VIP receptor. Expression of the VIP and PACAP receptors has been linked to the presence of human tumors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Pacap receptor; PACAP type II receptor; PACAP-R-2; PACAP-R2; pituitary adenylate cyclase activating polypeptide receptor, type II; Pituitary adenylate cyclase-activating polypeptide type II receptor; Pvr2; type 1 vasoactive intestinal peptide receptor; Vasoactive intestinal polypeptide receptor 1; VIP and PACAP receptor 1; VIP receptor 1; VIP receptor subtype 1; VIP receptor, type I; VIP-R-1; VIP1 receptor; VPAC1; VPAC1 receptor
Gene Aliases: AV071699; HVR1; II; PACAP-R-2; PACAP-R2; RDC1; V1RG; VAPC1; VIP-R-1; VIP-R1; VIPR; VIPR1; VIRG; VPAC1; VPAC1R; VPCAP1R
UniProt ID: (Human) P32241, (Mouse) P97751
Entrez Gene ID: (Human) 7433, (Mouse) 22354
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support