Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MRTLLPPALL TCWLLAPVNS IHPECRFHLE IQEEETKCAE LLRSQTEKHK ACSGVWDNIT CWRPANVGET VTVPCPKVFS NFYSKAGNIS KNCTSDGWSE TFPDFVDACG YSDPEDESKI TFYILVKAIY TLGYSVSLMS LATGSIILCL FRKLHCTRNY IHLNLFLSFI LRAISVLVKD DVLYSSSGTL HCPDQPSSWV GCKLSLVFLQ YCIMANFFWL LVEGLYLHTL LVAMLPPRRC FLAYLLIGWG LPTVCIGAWT AARLYLEDTG CWDTNDHSVP WWVIRIPILI SIIVNFVLFI SIIRILLQKL TSPDVGGNDQ SQYKRLAKST LLLIPLFGVH YMVFAVFPIS ISSKYQILFE LCLGSFQGLV VAVLYCFLNS EVQCELKRKW RSRCPTPSAS RDYRVCGSSF SRNGSEGALQ FHRGSRAQSF LQTETSVI
The Vasoactive Intestinal Polypeptide Receptor 2 (VPAC2) is a member of a family of three G-protein coupled receptors which mediate VIP (vasoactive intestinal peptide) and PACAP (pituitary adenlyate cyclase activating peptide) function in a variety of tissues. Together, the VPAC1 and VPAC2 receptors compose a subtype of the VIP receptor. Expression of the VIP and PACAP receptors has been linked to the presence of human tumors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Helodermin-preferring VIP receptor; PACAP type III receptor; Pacap type-3; PACAP3; PACAPR-3; Pituitary adenylate cyclase-activating polypeptide type III receptor; Pvr3; SCZD16; tcag7.970; Vasoactive intestinal polypeptide receptor 2; VIP and PACAP receptor 2; Vip type-2 receptor; VIP-R-2; Vip-r2; Vip2 receptor; VPAC2
Gene Aliases: C16DUPq36.3; DUP7q36.3; PACAP-R-3; PACAP-R3; VIP-R-2; VIP2R; VIPR2; VPAC2; VPAC2R; VPCAP2R
UniProt ID: (Human) P41587
Entrez Gene ID: (Human) 7434
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support