Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Keep as concentrated solution.
Predicted reactivity: Pig (84%), Rhesus Monkey (92%).
Positive Control: Target recombinant protein.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
VMO1 (Vitelline Membrane Outer Layer 1 Homolog) is a Protein Coding gene.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ERGA6350 antibody; MGC125880 antibody; MGC125881 antibody; PRO21055 antibody; vitelline membrane outer layer 1 homolog (chicken) antibody; Vitelline membrane outer layer protein 1 homolog; vitelline membrane outer layer protein 1 homolog antibody; VMO1 antibody
Gene Aliases: ERGA6350; Gm741; PRO21055; UNQ6350/PRO21055; VMO1
UniProt ID: (Human) Q7Z5L0, (Mouse) Q5SXG7
Entrez Gene ID: (Human) 284013, (Mouse) 327956
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support