Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: STLCYFTRS PPSLHMSLFP NPSWWLLNTS ALITACFPMV SPFVLMSRHP RIPRLGSACC GRNPQFPKLV R (242-311 aa encoded by BC107074)
Putative pheromone receptor. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC129709; MGC129710; V1r-like receptor 3; Vomeronasal type-1 receptor 3
Gene Aliases: FKSG46; V1RL3; V1RL3p; VN1R3
UniProt ID: (Human) Q9BXE9
Entrez Gene ID: (Human) 317702
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support