Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: EIHLSPGKF EVLKDGRLVN KNGSSGPILT VSLFGRWYTK DSLYSSCGTS NSAITYLLIF ILLMIIALQN IVML (448-520 aa encoded by B C126145)
This gene product is a member of the Vanin family of proteins that share extensive sequence similarity with each other, and also with biotinidase. The family includes secreted and membrane-associated proteins, a few of which have been reported to participate in hematopoietic cell trafficking. No biotinidase activity has been demonstrated for any of the vanin proteins, however, they possess pantetheinase activity, which may play a role in oxidative-stress response. The encoded protein is a GPI-anchored cell surface molecule that plays a role in transendothelial migration of neutrophils. This gene lies in close proximity to, and in same transcriptional orientation as two other vanin genes on chromosome 6q23-q24. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Glycosylphosphatidyl inositol-anchored protein GPI-80; pantetheinase; Pantetheine hydrolase VNN2; Protein FOAP-4; Vanin-2; Vannin 2; Vascular non-inflammatory molecule 2
Gene Aliases: FOAP-4; GPI-80; VNN2
UniProt ID: (Human) O95498
Entrez Gene ID: (Human) 8875
Molecular Function:
hydrolase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support