Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
Associates with the Ig-mu chain to form a molecular complex that is expressed on the surface of pre-B-cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: N27C7-2; pre-B lymphocyte gene 3; Pre-B lymphocyte protein 3; Protein VPreB3
Gene Aliases: 8HS20; N27C7-2; UNQ355/PRO619; VPREB3
UniProt ID: (Human) Q9UKI3
Entrez Gene ID: (Human) 29802
Molecular Function: immunoglobulin
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support