Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Positive Samples: HeLa, Mouse lung, Mouse brain, Rat brain
Immunogen sequence: MASILDEYEN SLSRSAVLQP GCPSVGIPHS GYVNAQLEKE VPIFTKQRID FTPSERITSL VVSSNQLCMS LGKDTLLRID LGKANEPNHV ELGRKDDAKV
VPS18 is a vesicle mediated protein that plays an important role in segregation of intracellular molecules into distinct organelles. Genetic studies in yeast have identified more than 40 vacuolar protein sorting (VPS) genes involved in vesicle transport to vacuoles. This gene encodes the human homolog of yeast class C Vps18 protein. The mammalian class C Vps proteins are predominantly associated with late endosomes/lysosomes, and like their yeast counterparts, may mediate vesicle trafficking steps in the endosome/lysosome pathway.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hVPS18; vacuolar protein sorting 18 homolog; vacuolar protein sorting protein 18; Vacuolar protein sorting-associated protein 18 homolog; VPS18 CORVET/HOPS core subunit
Gene Aliases: 9930024E13Rik; KIAA1475; PEP3; VPS18
UniProt ID: (Human) Q9H268, (Mouse) Q8R307
Entrez Gene ID: (Human) 57617, (Mouse) 228545, (Rat) 296083
Molecular Function: membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support