Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Immunogen sequence: MAMSFEWPWQ YRFPPFFTLQ PNVDTRQKQL AAWCSLVLSF CRLHKQSSMT VMEAQESPLF NNVKLQRKLP VESIQIVLEE LRKKGNLEWL DKSKSSFLIM WRRPEEWGKL IYQWVSRSGQ NNSVFTLYEL TNGEDTEDEE FHGLDEATLL RALQALQQEH KAEIITVSDG RGVKFF; Positive Samples: BxPC-3, Mouse brain, Mouse heart, Rat liver, Rat lung; Cellular Location: Cytoplasm, Endosome membrane, Nucleus, nucleoplasm
VPS25 (Vacuolar Protein Sorting 25 Homolog) is a Protein Coding gene. Among its related pathways are Delta508-CFTR traffic / ER-to-Golgi in CF and Budding and maturation of HIV virion. Gene Ontology (GO) annotations related to this gene include protein homodimerization activity and protein N-terminus binding.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dermal papilla-derived protein 9; ELL-associated protein of 20 kDa; ESCRT-II complex subunit VPS25; HGNC:28122; hVps25; MGC10540; Vacuolar protein-sorting-associated protein 25
Gene Aliases: 1110020N13Rik; AW107467; D11Wsu68e; DERP9; EAP20; FAP20; VPS25
UniProt ID: (Human) Q9BRG1, (Rat) P0C0A1, (Mouse) Q9CQ80
Entrez Gene ID: (Human) 84313, (Rat) 681059, (Mouse) 28084
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support