Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
This gene encodes a protein involved in endosomal sorting of cell surface receptors via a multivesicular body/late endosome pathway. The encoded protein is one of the three subunits of the ESCRT-I complex (endosomal complexes required for transport) involved in the sorting of ubiquitinated proteins. The two other subunits of ESCRT-I are vesicular protein sorting 23, also known as tumor susceptibility gene 101 (TSG101), and vesicular protein sorting 37. Two alternative transcripts encoding different isoforms have been described. Additional alternative transcripts may exist but the proteins encoded by these transcripts have not been verified experimentally.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Caspase-activated DNase inhibitor that interacts with ASK1; CIIA; ESCRT-I complex subunit VPS28; H-Vps28; MGC60323; vacuolar protein sorting 28 homolog; vacuolar protein sorting 28-like protein; Vacuolar protein sorting-associated protein 28 homolog; VPS28, ESCRT-I subunit; yeast class E protein Vps28p homolog
Gene Aliases: 1110014J03Rik; CIIA; D730005C08Rik; VPS28
UniProt ID: (Human) Q9UK41, (Mouse) Q9D1C8
Entrez Gene ID: (Dog) 607677, (Human) 51160, (Mouse) 66914, (Rat) 300052
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support