Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
This gene belongs to a group of vacuolar protein sorting genes that, when functionally impaired, disrupt the efficient delivery of vacuolar hydrolases. The protein encoded by this gene is a component of a large multimeric complex, termed the retromer complex, which is involved in retrograde transport of proteins from endosomes to the trans-Golgi network. This VPS protein may be involved in the formation of the inner shell of the retromer coat for retrograde vesicles leaving the prevacuolar compartment. Alternative splice variants encoding different isoforms, and usage of multiple polyadenylation sites have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp564F0223; FLJ20492; hVPS29; PEP11 homolog; retromer protein; vacuolar protein sorting 29 homolog; Vacuolar protein sorting-associated protein 29; vacuolar sorting protein 29; vacuolar sorting protein VPS29/PEP11; Vesicle protein sorting 29; VPS29 retromer complex component; x 007 protein
Gene Aliases: 2010015D08Rik; AW049835; DC15; DC7; MDS007; PEP11; VPS29
UniProt ID: (Human) Q9UBQ0, (Mouse) Q9QZ88
Entrez Gene ID: (Human) 51699, (Mouse) 56433
Molecular Function:
membrane traffic protein
vesicle coat protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support