Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibody detects endogenous levels of total VPS33A.
Vesicle mediated protein sorting plays an important role in segregation of intracellular molecules into distinct organelles. Genetic studies in yeast have identified more than 40 vacuolar protein sorting (VPS) genes involved in vesicle transport to vacuoles. This gene is a member of the Sec-1 domain family, and it encodes a protein similar to the yeast class C Vps33 protein. The mammalian class C VPS proteins are predominantly associated with late endosomes/lysosomes, and like their yeast counterparts, may mediate vesicle trafficking steps in the endosome/lysosome pathway.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: buff; hVPS33A; vacuolar protein sorting 33 homolog A; vacuolar protein sorting 33A; vacuolar protein sorting protein 33a; Vacuolar protein sorting-associated protein 33A; vacuolar sorting protein 33a; VPS33A CORVET/HOPS core subunit
Gene Aliases: 3830421M04Rik; AI503300; AW048546; AW554476; Bf; Buff; r-Vps33a; VPS33A
UniProt ID: (Human) Q96AX1, (Mouse) Q9D2N9
Entrez Gene ID: (Human) 65082, (Mouse) 77573, (Rat) 65081
Molecular Function:
membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support