Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSSTSPNLQ KAIDLASKAA QEDKAGNYEE ALQLYQHAVQ YFLHVVKYEA QGDKAKQSIR AKCTEYLDRA EKLKEYLKNK EKKAQKPVKE GQPSPADEKG NDSDGEGESD DPEKKKLQNQ LQGAIVIERP NVKWSDVAGL EGAKEALKEA VILPIKFPHL FTGKRTPWRG ILLFGPPGTG KSYLAKAVAT EANNSTFFSI SSSDLVSKWL GESEKLVKNL FQLARENKPS IIFIDEIDSL CGSRSENESE AARRIKTEFL VQMQGVGVDN DGILVLGATN IPWVLDSAIR RRFEKRIYIP LPEPHARAAM FKLHLGTTQN SLTEADFREL GRKTDGYSGA DISIIVRDAL MQPVRKVQSA THFKKVRGPS RADPNHLVDD LLTPCSPGDP GAIEMTWMDV PGDKLLEPVV SMSDMLRSLS NTKPTVNEHD LLKLKKFTED FGQEG (1-444 aa encoded by BC039574)
The protein encoded by this gene is a member of the AAA protein family (ATPases associated with diverse cellular activities), and is the homolog of the yeast Vps4 protein. In humans, two paralogs of the yeast protein have been identified. The former share a high degree of aa sequence similarity with each other, and also with yeast Vps4 and mouse Skd1 proteins. Mouse Skd1 (suppressor of K+ transport defect 1) has been shown to be a yeast Vps4 ortholog. Functional studies indicate that both human paralogs associate with the endosomal compartments, and are involved in intracellular protein trafficking, similar to Vps4 protein in yeast. The gene encoding this paralog has been mapped to chromosome 18; the gene for the other resides on chromosome 16.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cell migration-inducing 1; Cell migration-inducing gene 1 protein; Protein SKD1; Suppressor of K(+) transport growth defect 1; suppressor of K+ transport defect 1; vacuolar protein sorting 4B; Vacuolar protein sorting-associated protein 4B
Gene Aliases: 8030489C12Rik; MIG1; SKD1; SKD1B; VPS4-2; VPS42; VPS4B
UniProt ID: (Human) O75351, (Mouse) P46467
Entrez Gene ID: (Human) 9525, (Mouse) 20479, (Rat) 360834
Molecular Function:
non-motor microtubule binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support