Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
CTXL, a cortical thymocyte marker, is a member of the immunoglobulin (Ig) superfamily and has features of both antigen specific receptors and adhesion molecules. This single pass, type I protein contains a signal peptide, an extracellular V type Ig like domain followed by a C2 type Ig like domain and a cytoplasmic tail. Expression levels of CTXL are high in the stomach, colon, prostate, trachea, and thyroid gland, with lower levels observed in bladder and lung tissues. CTXL is not expressed in the thymus. CTXL is developmentally regulated and may be involved in thymocyte selection. It localizes to junctional complexes between endothelial and epithelial cells and may play a role in cell-cell adhesion and transmigration of leukocytes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cortical thymocyte receptor (X. laevis CTX) like; Cortical thymocyte-like protein; CT-like protein; V-set and immunoglobulin domain-containing protein 2
Gene Aliases: 2210413P10Rik; CTH; CTXL; UNQ2770/PRO7154; VSIG2
UniProt ID: (Human) Q96IQ7
Entrez Gene ID: (Human) 23584
Molecular Function:
defense/immunity protein
immunoglobulin receptor superfamily
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support