Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
T cell activation by APCs is positively and negatively regulated by members of the B7 family. VSIG4 is a strong negative regulator of murine and human T cell proliferation and IL-2 production.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: complement receptor of the immunoglobulin superfamily; Ig superfamily protein; Protein Z39Ig; V-set and immunoglobulin domain-containing protein 4
Gene Aliases: CRIg; UNQ317/PRO362; VSIG4; Z39IG
UniProt ID: (Human) Q9Y279
Entrez Gene ID: (Human) 11326
Molecular Function: defense/immunity protein immunoglobulin receptor superfamily
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support