Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Positive control: Human liver tissue lysate, Hela cell lysate, mouse brain tissue lysate, rat brain tissue lysate, rat cerebellum tissue, human brain tissue, mouse brain tissue.
Predicted band size: 22 kDa
Subcellular Location: Cytosol, membrane.
This gene is a member of the visinin/recoverin subfamily of neuronal calcium sensor proteins. The encoded protein is strongly expressed in granule cells of the cerebellum where it associates with membranes in a calcium-dependent manner and modulates intracellular signaling pathways of the central nervous system by directly or indirectly regulating the activity of adenylyl cyclase. Alternatively spliced transcript variants have been observed, but their full-length nature has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 21 kDa CABP; Hippocalcin-like protein 3; HLP3; Neural visinin-like protein 1; neurocalcin alpha; NVL-1; NVP-1; VILIP; visinin-like 1; Visinin-like protein 1; VLP-1
Gene Aliases: HLP3; HPCAL3; HUVISL1; Nvp1; Ratnvp1; VILIP; VILIP-1; VISL1; Vnsl1; VSNL1
UniProt ID: (Human) P62760, (Mouse) P62761
Entrez Gene ID: (Human) 7447, (Mouse) 26950, (Rat) 24877
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support