Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product is supplied at 1 mL (100 µL/mL).
Application Notes: Western blot - non-reduced sample treatment and SDS-Page was used. The band sizes are 45 kDa and fainter band of 37 kDa in transfected cells and 37 kDa in granulocytes. Functional Assay - Antibody 1A5 showed inhibition of Fc eta RI-mediated degranulation by RBL-2H3 cells transfected with SIRL-1. The percentage of inhibition of degranulation by SIRL-1 was calculated by (OD405 TNP stimulation - OD405 TNP with SIRL-1 crosslinking)/(OD405 TNP stimulation - OD405 spontaneous release) x 100.
VSTM1 (V-Set And Transmembrane Domain Containing 1) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include cytokine activity. An important paralog of this gene is LILRB1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: LAIR homolog; OSCAR-like transcript-1; Signal inhibitory receptor on leukocytes-1; SIRL-1; V-set and transmembrane domain-containing protein 1
Gene Aliases: SIRL-1; SIRL1; UNQ3033; UNQ3033/PRO9835; VSTM1
UniProt ID: (Human) Q6UX27
Entrez Gene ID: (Human) 284415
Molecular Function:
immunoglobulin receptor superfamily
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support