Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Sequence of this protein is as follows: MGAPLAVALG ALHYLALFLQ LGGATRPAGH APWDNHVSGH ALFTETPHDM TARTGEDVEM ACSFRGSGSP SYSLEIQWWY VRSHRDWTDK QAWASNQLKA SQQEDAGKEA TKISVVKVVG SNISHKLRLS RVKPTDEGTY ECRVIDFSDG KARHHKVKAY LRVQPGENSV LHLPEAPPAA PAPPPPKPGK ELRKRSVDQE ACSL
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: V-set and transmembrane domain-containing protein 2-like protein
Gene Aliases: C20orf102; dJ1118M15.2; VSTM2L
UniProt ID: (Human) Q96N03
Entrez Gene ID: (Human) 128434
Molecular Function:
immunoglobulin receptor superfamily
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support