Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSSDFEGYE QDFAVLTAEI TSKIARVPRL PPDEKKQMVA NVEKQLEEAK ELLEQMDLEV REIPPQSRGM YSNRMRSYKQ EMGKLETDFK RSRIAYSDEV RNELLGDDGN SSENQRAHLL DNTERLERSS RRLEAGYQIA VETEQIGQEM LENLSHDREK IQRARERLRE TDANLGKSSR ILTGMLRRGC SVKKQCNLSL APKA (1-203 aa encoded by BC017052)
V-SNARE that mediates vesicle transport pathways through interactions with t-SNAREs on the target membrane. These interactions are proposed to mediate aspects of the specificity of vesicle trafficking and to promote fusion of the lipid bilayers. Involved in vesicular transport from the late endosomes to the trans-Golgi network. Along with VAMP7, involved in an non- conventional RAB1-dependent traffic route to the cell surface used by KCNIP1 and KCND2. May be involved in increased cytokine secretion associated with cellular senescence.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: SNARE Vti1a-beta protein; vesicle transport through interaction with t-SNAREs 1 homolog; Vesicle transport through interaction with t-SNAREs homolog 1A; Vesicle transport v-SNARE protein Vti1-like 2; Vti1-rp2
Gene Aliases: 1110014F16Rik; 1110018K19Rik; 4921537J05Rik; MMDS3; MVti1; MVti1a; Vti1; Vti1-rp2; VTI1A; Vti1l2; VTI1RP2
UniProt ID: (Human) Q96AJ9, (Mouse) O89116, (Rat) Q9JI51
Entrez Gene ID: (Human) 143187, (Mouse) 53611, (Rat) 65277
Molecular Function:
SNARE protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support