Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Dilutions may be prepared by adding phosphate buffered saline (PBS, pH 7.2). Repeated thawing and freezing should be avoided. If a slight precipitation occurs upon storage, this should be removed by centrifugation. It will not affect the performance of the antiserum. Diluted antiserum should be stored at 4 C, not refrozen, and preferably used the same day.
In immunoelectrophoresis use 2 ul serum or equivalent against 120 ul antiserum. In double radial immunodiffusion use a rosette arrangement with 10 ul antiserum in 3 mm diameter centre well and 2 ul serum samples (neat and serially diluted) in 2 mm diameter peripheral wells.
VAV2 is the second member of the VAV guanine nucleotide exchange factor family of oncogenes. Unlike VAV1, which is expressed exclusively in hematopoietic cells, VAV2 transcripts were found in most tissues. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Guanine nucleotide exchange factor VAV2; Oncogene VAV2; vav 2 guanine nucleotide exchange factor; vav 2 oncogene; VAV-2
Gene Aliases: VAV-2; VAV2
UniProt ID: (Human) P52735
Entrez Gene ID: (Human) 7410
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support