Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Exalpha
Precautions should be taken upon storage for longer periods. Problems of long term stability may occur with highly diluted solutions. No other preservative agent has been added to the present formulation. For long storage purposes in solution the addition of sodium azide at 0.02 % is advised with the appropriate precautions of use.
The antibody may be used in ELISA and IAC. Depending on conjugate: dilution 1/75,000 . (The titer is defined as the dilution that gives 50 % of the absorbance from the maximum absorbance when tested with ELISA). Suggested concentration to use in ELISA: 1/75,000 from the delivered solution. Plates are coated with 400 ng/ml BSA-conjugated C6-alkylphenol. HRP-conjugated anti-rabbit IgG as a tracer 1/8,000
Vesicle-associated membrane protein 2 is essential for cAMP-regulated exocytosis. VAMP (also know as synaptobrevin) is part of the synaptic vesicle docking and fusion complex and plays a central role in neuroexocytosis. Two VAMP (vesicle-associated membrane protein) isoforms are expressed in the nervous system and are differently distributed among the specialized parts of the tissue. VAMP-1 and -2 are present in all tissues tested, including kidney, adrenal gland, liver, pancreas, thyroid, heart, and smooth muscle. The two isoforms are differentially expressed in various tissues and their level may depend on differentiation. VAMP-1 is restricted to exocrine pancreas and to kidney tubular cells, whereas VAMP-2 is the predominant isoform present in Langerhans islets and in glomerular cells. Both isoforms show a patchy vesicular intracellular distribution in confocal microscopy. Evidence for the importance of neuronal VAMPproteins in the physiology of all cells is indicated.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: synaptobrevin 1; synaptobrevin 2; Synaptobrevin 2 (vesicle-associated membrane protein VAMP-2); Synaptobrevin-1; Synaptobrevin-2; VAMP-1; VAMP-2; Vesicle-associated membrane protein (synaptobrevin 2); Vesicle-associated membrane protein 1; vesicle-associated membrane protein 1 (synaptobrevin 1); Vesicle-associated membrane protein 2; vesicle-associated membrane protein 2 (synaptobrevin 2)
Gene Aliases: RATVAMPB; RATVAMPIR; SPAX1; SYB; SYB1; SYB2; VAMP-1; VAMP-2; VAMP1; VAMP2
UniProt ID: (Human) P23763, (Human) P63027, (Rat) Q63666, (Rat) P63045
Entrez Gene ID: (Human) 6843, (Human) 6844, (Rat) 25624, (Rat) 24803
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support