Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MTKLSAQVK GSLNITTPGL QIWRIEAMQM VPVPSSTFGS FFDGDCYIIL AIHKTASSLS YDIHYWIGQD SSLDEQGAAA IYTTQMDDFL KGRAVQHREV QGNESEAFRG YFKQGLVIRK GGVASGMKHV ETNSYDVQRL LHVKGKRNVV AGEVEMSWKS FNRGDVFLLD LGKLIIQWNG PESTRMERLR GMTLAKEIRD QERGGRTYVG VVDGENELAS PKLMEVMNHV LGKRRELKAA VPDTVVEPAL KAALKLYHVS DSEGNLVVRE VATRPLTQDL LSHEDCYILD QGGLKIYVWK GKKANEQEKK GAMSHALNFI KAKQYPPSTQ VEVQNDGAES AVFQQLFQKW T (1-350 aa encoded by BC017303)
Villin is part of cytoskeleton associated with the microvillar actin core bundle of gastrointestinal and renal brush border. It can interact with actin in a Ca2+ and phosphoinositide-regulated manner. Villin is a very specific marker for adenocarcinomas of the gastrointestinal tract and the pancreas. It is also expressed in some Merkel cell tumor and adenocarcinoma of the lung, prostate, ovarian and kidney.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: OTTHUMP00000207159; Villin-1
Gene Aliases: D2S1471; VIL; VIL1
UniProt ID: (Human) P09327, (Mouse) Q62468
Entrez Gene ID: (Human) 7429, (Mouse) 22349
Molecular Function:
non-motor actin binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support