Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reactivity: MW 57,000 intermediate filament protein (vimentin) of mesenchymal cells. Tumors specifically detected: sarcoma (including myosarcoma), lymphoma, melanoma. Reactivity on cultured cell lines: fibroblasts (SV-80).
Vimentin is a developmentally regulated intermediate filament protein (IFP) found in cells of mesenchymal origin. It is believed to be involved with the intracellular transport of proteins between the nucleus and plasma membrane. Unlike other IFP proteins, vimentin is expressed, along with desmin, during the early stages of cellular development. During the development process, vimentin is exchanged for new, tissue-specific IFPs. Vimentin has been implicated to be involved in the rate of steroid synthesis via its role as a storage network for steroidogenic cholesterol containing lipid droplets. Vimentin phosphorylation by a protein kinase causes the breakdown of intermediate filaments and activation of an ATP and myosin light chain dependent contractile event. This results in cytoskeletal changes that facilitate the interaction of the lipid droplets within mitochondria, and subsequent transport of cholesterol to the organelles leading to an increase in steroid synthesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: epididymis luminal protein 113; FLJ36605; RP11-124N14.1; VIME; Vimentin; vimentin C-terminus
Gene Aliases: CTRCT30; HEL113; VIM
UniProt ID: (Bovine) P48616, (Human) Q6LER9
Entrez Gene ID: (Bovine) 280955, (Chicken) 420519, (Human) 7431
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support