Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: HepG2.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The SORBS3 gene encodes 2 isoforms vinexin-alpha and -beta that are vinculin (VCL; MIM 193065)-binding cytoskeletal proteins involved in focal adhesion and cell-cell adhesion (Kioka et al., 1999 [PubMed 9885244]).
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: SCAM-1; SH3-containing adapter molecule 1; Sorbin and SH3 domain-containing protein 3; Vinexin; vinexin beta (SH3-containing adaptor molecule-1)
Gene Aliases: SCAM-1; SCAM1; SH3D4; SORBS3
UniProt ID: (Human) O60504
Entrez Gene ID: (Human) 10174
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support