Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: KLCMAALKH QPQEFPTYVE PTNDEICEAF RKDPKEYANQ FMWEYSTNYG QAPLSLLVSY TKSYLSMVGS CCTSASPTVC FLKERLQLKH LSLLTTLSNR VCSQYAAYGE KKSRLSNLIK LAQKVPTADL EDVLPLAEDI TNILSKCCES ASEDCMAKEL PEHTVKLCDN LSTKNSKFED CCQEKTAMDV FVCTYFMPAA QLPELPDVEL PTNKDVCDPG NTKVMDKYTF ELSRRTHLPE VFLSKVLEPT LKSLGECCDV EDSTTCFNAK GPLLKKELSS FIDKGQELCA DYSENTFTEY KKKLAERLKA KLPDATPTEL AKLVNKRSDF ASNCCSINSP PLYCDSEIDA ELKNIL (120-474 aa encoded by BC057228)
GC (vitamin D-binding protein) is involved in vitamin D transport/storage, scavenging of extracellular G-actin, enhancement of the chemotactic activity of C5 alpha for neutrophils in inflammation and macrophage activation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DBP; DBP-maf; DBP/ VDBG; epididymis secretory protein Li 51; Gc; Gc globulin; Gc protein-derived macrophage activating factor; Gc-globulin; Gc-MAF; Group specific component vitamin D binding protein; Group-specific component; group-specific component (vitamin D binding protein); hDBP; V VDB; VDB; Vitamin D binding alpha globulin; vitamin D-binding alpha-globulin; Vitamin D-binding protein; Vitamin D-binding protein-macrophage activating factor
Gene Aliases: DBP; DBP/GC; GC; Gc-MAF; GcMAF; GRD3; HEL-S-51; VDBG; VDBP
UniProt ID: (Human) P02774
Entrez Gene ID: (Human) 2638
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support