Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MALNKNHSE GGGVIVNNTE SILMSYDHVE LTFNDMKNVP EAFKGTKKGT VYLTPYRVIF LSKGKDAMQS FMMPFYLMKD CEIKQPVFGA NYIKGTVKAE AGGGWEGSAS YKLTFTAGGA IEFGQRMLQV ASQASRGEVP SGAYGYSYMP SGAYVYPPPV ANGMYPCPPG YPYPPPPPEF YPGPPMMDGA MGYVQPPPPP YPGPMEPPVS GPDVPSTPAA EAKAAEAAAS AYYNPGNPHN VYMPTSQPPP PPYYPPEDKK TQ (1-261 aa encoded by BC010616)
The globular WW domain is composed of 38 to 40 semiconserved amino acids shared by proteins of diverse functions including structural, regulatory, and signaling proteins. The domain is involved in mediating protein-protein interactions through the binding of polyproline ligands. WBP2 is a WW domain binding protein, which binds to the WW domain of Yes kinase-associated protein by its PY motifs. The function of this protein has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: WBP-2; WW domain-binding protein 2
Gene Aliases: GRAMD6; WBP-2; WBP2
UniProt ID: (Human) Q969T9
Entrez Gene ID: (Human) 23558
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support