Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LQVPETPAP EPAEVLEMQE DCPEAQIGQE AQDEDSSDSE ADEQSISQDP KDTPSQPSST SHRPRAASCR ATAAWCTDSG SDDSRRWSDQ LSLDEKDGFI FVNYSEGQTR AHLQGPLSHP HPNPIEVRNY SRLKPGYRWE RQLVFRSKLT MHTAFDRKDN AHPAEVTALG ISKDHSRILV GDSRGRVFSW SVSDQPGRSA ADHWVKDEGG DSCSGCSVRF SLTERRHHCR NCGQLFCQKC SRFQSEIKRL KISSPVRVCQ NCYYNLQHER GSEDGPRNC (3249-3526 aa encoded by BC013377)
The ubiquitously expressed cellular upstream stimulatory factor (USF) consists of 43kDa (USF-1) and 44kDa (USF-2) polypeptides which independently exhibit site-specific DNA binding and are members of the c-Myc-related family of regulatory factors containing helix-loop-helix domains. USF also contains a leucine repeat that is required for efficient DNA binding.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alfy; Autophagy-linked FYVE protein; WD repeat and FYVE domain-containing protein 3
Gene Aliases: ALFY; KIAA0993; WDFY3; ZFYVE25
UniProt ID: (Human) Q8IZQ1
Entrez Gene ID: (Human) 23001
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support