Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The ubiquitously expressed cellular upstream stimulatory factor (USF) consists of 43kDa (USF-1) and 44kDa (USF-2) polypeptides which independently exhibit site-specific DNA binding and are members of the c-Myc-related family of regulatory factors containing helix-loop-helix domains. USF also contains a leucine repeat that is required for efficient DNA binding.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alfy; Autophagy-linked FYVE protein; Beach domain, WD repeat and FYVE domain-containing protein 1; BWF1; galactosyltransferase 3 beta 1, 4; WD repeat and FYVE domain-containing protein 3
Gene Aliases: 2610509D04Rik; ALFY; AW319683; B930017C24; Bchs; BWF1; D5Ertd66e; Ggtb3; KIAA0993; mKIAA0993; WDFY3; ZFYVE25
UniProt ID: (Human) Q8IZQ1, (Mouse) Q6VNB8
Entrez Gene ID: (Human) 23001, (Mouse) 72145
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support