Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MGTSKEKAKT ATLKQALLGH TDTVTCATAS LAYHIIVSGS RDRTCIIWDL NKLSFLTQLR GHRAPVSALC INELTGDIVS CAGTYIHVWS INGNPIVSVN TFTGRSQQII CCCMSEMNEW DTQNVIVTGH SDGVVRFWRM EFLQVPETPA PEPAEVLEMQ EDCPEAQIGQ EAQDEDSSDS EADEQSISQD PKDTPSQPSS TSHRPRAASC RATAAWCTDS GSDDSRRWSD QLSLDEKDGF IFVNYSEGQT RAHLQGPLSH PHPNPIEVRN YSRLKPGYRW ERQLVFRSKL TMHTAFDRKD NAHPAEVTAL GISKDHSRIL VGDSRGRVFS WSVSDQPGRS AADHWVKDEG GDSCSGCSVR FSLTERRHHC RNCGQLFCQK CSRFQSEIKR LKISSPVRVC QNCYYNLQHE RGSEDGPRNC
The ubiquitously expressed cellular upstream stimulatory factor (USF) consists of 43kDa (USF-1) and 44kDa (USF-2) polypeptides which independently exhibit site-specific DNA binding and are members of the c-Myc-related family of regulatory factors containing helix-loop-helix domains. USF also contains a leucine repeat that is required for efficient DNA binding.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alfy; Autophagy-linked FYVE protein; WD repeat and FYVE domain-containing protein 3
Gene Aliases: ALFY; KIAA0993; WDFY3; ZFYVE25
UniProt ID: (Human) Q8IZQ1
Entrez Gene ID: (Human) 23001
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support