Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAGDTSLSE NYAFAGMYHV FDQHVDEAVP RVRFANDDRH RLACCSLDGS ISLCQLVPAP PTVLRVLRGH TRGVSDFAWS LSNDILVSTS LDATMRIWAS EDGRCIREIP DPDSAELLCC TFQPVNNNLT VVGNAKHNVH VMNISTGKKV KGGSSKLTGR VLALSFDAPG RLLWAGDDRG SVFSFLFDMA TGKLTKAKRL VVHEGSPVTS ISARSWVSRE ARDPSLLINA CLNKLLLYRV VDNEGTLQLK RSFPIEQSSH PVRSIFCPLM SFRQGACVVT GSEDMCVHFF DVERAAKAAV NKLQGHSAPV LDVSFNCDES LLASSDASGM VIVWRREQK (148-485 aa encoded by BC002507)
WDR13 is a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. WDR13 gene is widely expressed in various tissues, and located in chromosome X. The function of this gene has not been determined. This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. This gene is widely expressed in various tissues, and located in chromosome X. The function of this gene has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: memory-related protein; WD repeat-containing protein 13
Gene Aliases: 1700060B08Rik; 5730411P10Rik; DXHXS7467e; MG21; mMg21; RGD1560982; W51679; WDR13
UniProt ID: (Human) Q9H1Z4, (Mouse) Q91V09
Entrez Gene ID: (Human) 64743, (Rat) 317370, (Mouse) 73447
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support