Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MDNKISPEAQ VAELELDAVI GFNGHVPTGL KCHPDQEHMI YPLGCTVLIQ AINTKEQNFL QGHGNNVSCL AISRSGEYIA SGQVTFMGFK VDIILWDYKN RELLARLSLH KGKIEALAFS PNDLYLVSLG GPDDGSVVVW SIAKRDAICG SPAAGLNVGN ATNVIFSRCR DEMFMTAGNG TIRVWELDLP NRKIWPTESQ TGQLKRIVMS IGVDDDDSFF YLGTTTGDIL KMNPRTKLLT DVGPAKDKFS LGVSAIRCLK MGGLLVGSGA GLLVFCKSPG YKPIKKIQLQ GGITSITLRG EGHQFLVGTE ESHIYRVSFT DFKETLIATC HFDAVKDIVF PFGTAELFAT CAKKDIRVWH TSSNRELLRI TVPNMTCHGI DFMRDGKSII SAWNDGKIRA FAPETGRLMY VINNAHRIGV TAIATTSDCK RVISGGGEGE VRVWQIGCQT QKLEEALKEH KSSVSCIRVK RNNEECVTAS TDGTCIIWDL VRLRRNQMIL ANTLFQCVCY HPEEFQIITS GTDRKIAYWE VFDGTVIREL EGSLSGSING MDITQEGVHF VTGGNDHLVK VWDYNEGEVT HVGVGHSGNI TRIRISPGNQ YIVSVSADGA ILRWKYPYTS
WD repeat-containing proteins, such as WDR16, play crucial roles in a wide range of physiologic functions, including signal transduction, RNA processing, remodeling the cytoskeleton, regulation of vesicular traffic, and cell division (Silva et al., 2005 ).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cilia- and flagella-associated protein 52; WD repeat domain 16; WD repeat-containing protein 16; WD40-repeat protein up-regulated in HCC; WD40-repeat protein upregulated in HCC
Gene Aliases: CFAP52; WDR16; WDRPUH
UniProt ID: (Human) Q8N1V2
Entrez Gene ID: (Human) 146845
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support