Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: GTPPLLCGKV SRDIRQEIEK LTANSQVKKL RWFSECLSPP GGSDNLWNLV AVIKGQDDSL LPQNYCKGIM HLKHLIKFRT SEAQELTTVK M
This gene encodes a WD repeat-containing protein. It is abundantly expressed in retina and testis, and is thought to be a candidate gene for retinal disease. Alternatively spliced transcript variants have been found for this gene. [provided by RefSeq, Nov 2009].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: WD repeat-containing protein 17
Gene Aliases: WDR17
UniProt ID: (Human) Q8IZU2
Entrez Gene ID: (Human) 116966
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support