Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MKRIFSLLE KTWLGAPIQF AWQKTSGNYL AVTGADYIVK IFDRHGQKRS EINLPGNCVA MDWDKDGDVL AVIAEKSSCI YLWDANTNKT SQLDNGMRDQ MSFLLWSKVG SFLAVGTVKG NLLIYNHQTS RKIPVLGKHT KRITCGCWNA ENLLALGGED KMITVSNQEG DTIRQTQVRS EPSNMQFFLM KMDDRTSAAE SMISVVLGKK TLFFLNLNEP DNPADLEFQQ DFGNIVCYNW YGDGRIMIGF SCGHFVVIST HTGELGQEIF QARNHKDNLT SIAVSQTLNK VATCGDNCIK IQDLVDLKDM YVILNLDEEN KGLGTLSWTD DGQLLALSTQ RGSLHVFLTK LPIL (1-353 aa encoded by BC032578)
This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. This protein contains six WD repeats, a clathrin heavy-chain repeat, and three transmembrane domains. This gene is conserved from C. elegans to human. It may participate in androgen-regulated signaling mechanisms or in the vesicular trafficking of androgen-regulated secretory processes. Alternatively spliced transcript variants encoding distinct isoforms have been reported but the full-length nature of one of these variants has not been defined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Intraflagellar transport 144 homolog; WD repeat membrane protein PWDMP; WD repeat-containing protein 19
Gene Aliases: ATD5; C330027H04Rik; CED4; D330023L08Rik; DYF-2; DYF2; IFT144; KIAA1638; mKIAA1638; NPHP13; ORF26; Oseg6; PWDMP; RGD1306997; SRTD5; WDR19
UniProt ID: (Human) Q8NEZ3, (Mouse) Q3UGF1
Entrez Gene ID: (Human) 57728, (Rat) 305349, (Mouse) 213081
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support