Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: NSKDQTIKLW DIRRFSSREG MEASRQAATQ QNWDYRWQQV PKKAWRKLKL PGDSSLMTYR GHGVLHTLIR CRFSPIHSTG QQFIYSGCST GKVVVYDLLS GHIVKKLTNH KACVRDVSWH PFEEKIVSSS WDG
Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 98%.
WDR23 contains 7 WD repeats. The function of WDR23 remains unknown.This gene encodes a WD repeat-containing protein. Multiple alternatively spliced transcript variants have been found for this gene, but the full-length nature of some variants has not been defined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DDB1- and CUL4-associated factor 11; DKFZp779A1629; WD repeat domain 23; WD repeat-containing protein 23
Gene Aliases: DCAF11; GL014; PRO2389; WDR23
UniProt ID: (Human) Q8TEB1
Entrez Gene ID: (Human) 80344
Molecular Function:
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support